Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012528.1 | NP_036660.1 | 2, 3 | 272 | ||
GenBank mRNA | X74833.1 | - | 2, 3 | 272 |