Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ace | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012544.1 | NP_036676.1 | 9, 10 | 1524 | ||
GenBank mRNA | AF201331.1 | - | 9, 10 | 1524 | ||
AF201332.1 | - | 9, 10 | 1524 | |||
AF532783.1 | - | 9, 10 | 1468 | |||
AF532784.1 | - | 9, 10 | 1468 | |||
BC085760.1 | - | 9, 10 | 1524 | 7097136 | ||
U03708.1 | - | 9, 10 | 1524 | |||
U03734.1 | - | 9, 10 | 1524 |