Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prl8a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022846.2 | NP_074037.1 | 6 | 681 | ||
XM_006253906.1 | XP_006253968.1 | 6 | 741 | |||
GenBank mRNA | BC097316.1 | - | 6 | 682 | 7442457 | |
FQ219955.1 | - | 6 | 741 | |||
FQ220067.1 | - | 6 | 740 | |||
FQ220110.1 | - | 6 | 741 | |||
FQ222092.1 | - | 2 | 227 | |||
FQ222955.1 | - | 6 | 741 | |||
FQ228468.1 | - | 2 | 311 | |||
FQ229664.1 | - | 6 | 715 | |||
FQ229702.1 | - | 6 | 739 | |||
FQ229765.1 | - | 6 | 737 | |||
L06441.1 | - | 6 | 683 |