Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Drd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012547.1 | NP_036679.1 | 7 | 1412 | ||
XM_006242979.2 | XP_006243041.1 | 6 | 1106 | |||
XM_008766190.1 | XP_008764412.1 | 6 | 1429 | |||
XM_008766191.1 | XP_008764413.1 | 5 | 1342 | |||
GenBank mRNA | M36831.1 | - | 6 | 1106 | ||
X53278.1 | - | 7 | 1192 | |||
X56065.1 | - | 7 | 1412 |
Set Membership: |
![]() |