Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGCG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000231.2 | NP_000222.1 | 4, 5 | 537 | ||
XM_005266505.2 | XP_005266562.1 | 5, 6 | 630 | |||
XM_006719861.3 | XP_006719924.1 | 4, 5 | 721 | |||
GenBank mRNA | AK290620.1 | - | 5, 6 | 521 | ||
AK316579.1 | - | 5, 6 | 605 | |||
BC074777.2 | - | 5, 6 | 425 | 30915556 | ||
BC074778.2 | - | 5, 6 | 424 | 30915334 | ||
BC109321.2 | - | 4, 5 | 497 | 40036813 | ||
DA897096.1 | - | 4, 5 | 453 | |||
U34976.1 | - | 5, 6 | 506 |