Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012568.2 | NP_036700.1 | 8, 9 | 1624 | ||
XM_003752039.3 | XP_003752087.1 | 9, 10 | 1248 | |||
XM_008773152.1 | XP_008771374.1 | 8, 9 | 1617 | |||
XM_008773153.1 | XP_008771375.1 | 8, 9 | 1030 | |||
GenBank mRNA | AJ310837.1 | - | 8, 9 | 1073 | ||
X61159.1 | - | 8, 9 | 1624 |