Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HBB SIRNA Assays ›
Gene Symbol:
Hbb
Gene Name:
hemoglobin, beta
Gene Aliases:
-
Chromosome Location:
Chr.1: 168971269 - 168972680 on Build Rnor_6.0
UniGene ID:
Rn.202945
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s127870
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Hbb
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_033234.1
NP_150237.1
2
175
GenBank mRNA
BC058448.1
-
2
184
6921704
FQ209466.1
-
2
180
FQ209544.1
-
2
180
FQ209725.1
-
2
182
FQ209747.1
-
2
180
FQ209754.1
-
2
180
FQ209760.1
-
2
180
FQ209781.1
-
2
180
FQ209789.1
-
2
180
FQ209796.1
-
2
180
FQ209806.1
-
2
182
FQ209822.1
-
180
FQ209826.1
-
2
189
FQ209841.1
-
2
182
FQ209870.1
-
2
182
FQ209871.1
-
2
180
FQ209903.1
-
2
180
FQ209929.1
-
2
180
FQ209937.1
-
2
180
FQ209961.1
-
2
179
FQ209989.1
-
2
180
FQ209996.1
-
2
180
FQ210038.1
-
2
182
FQ210054.1
-
2
180
FQ210058.1
-
2
180
FQ210090.1
-
2
182
FQ210123.1
-
2
180
FQ210127.1
-
2
180
FQ210137.1
-
2
180
FQ210175.1
-
2
178
FQ210181.1
-
2
182
FQ210200.1
-
2
180
FQ210216.1
-
2
179
FQ210218.1
-
2
180
FQ210280.1
-
2
203
FQ210343.1
-
2
180
FQ210348.1
-
2
180
FQ210353.1
-
2
180
FQ210373.1
-
2
180
FQ210415.1
-
2
180
FQ210449.1
-
2
180
FQ210466.1
-
2
180
FQ210482.1
-
2
180
FQ210486.1
-
2
182
FQ210495.1
-
2
180
FQ210497.1
-
2
181
FQ210499.1
-
2
180
FQ210511.1
-
2
180
FQ210523.1
-
2
180
FQ210534.1
-
2
180
FQ210602.1
-
2
180
FQ210676.1
-
2
180
FQ210677.1
-
2
180
FQ210735.1
-
2
179
FQ210771.1
-
2
180
FQ210783.1
-
2
172
FQ210789.1
-
2
180
FQ210794.1
-
2
180
FQ210798.1
-
2
180
FQ210806.1
-
2
180
FQ210826.1
-
2
180
FQ210828.1
-
2
177
FQ210829.1
-
2
180
FQ210838.1
-
2
180
FQ210852.1
-
2
180
FQ210875.1
-
2
180
FQ210912.1
-
2
180
FQ210937.1
-
2
180
FQ210949.1
-
2
183
FQ210960.1
-
2
180
FQ210984.1
-
2
180
FQ210985.1
-
2
180
FQ210998.1
-
2
180
FQ211014.1
-
2
180
FQ211016.1
-
2
180
FQ211034.1
-
2
180
FQ211061.1
-
2
179
FQ211065.1
-
2
180
FQ211103.1
-
2
180
FQ211125.1
-
2
182
FQ211133.1
-
2
177
FQ211137.1
-
2
180
FQ211140.1
-
2
180
FQ211142.1
-
2
182
FQ211147.1
-
2
180
FQ211154.1
-
2
180
FQ211182.1
-
2
180
FQ211193.1
-
2
180
FQ211207.1
-
2
182
FQ211241.1
-
2
180
FQ211242.1
-
2
180
FQ211290.1
-
2
180
FQ211314.1
-
2
180
FQ211331.1
-
2
180
FQ211336.1
-
2
180
FQ211338.1
-
2
179
FQ211340.1
-
2
180
FQ211423.1
-
2
182
FQ211443.1
-
2
180
FQ211452.1
-
2
180
FQ211477.1
-
2
180
FQ211497.1
-
2
180
FQ211548.1
-
2
180
FQ211583.1
-
2
180
FQ211590.1
-
2
181
FQ211613.1
-
2
182
FQ211617.1
-
2
180
FQ211626.1
-
2
180
FQ211692.1
-
2
180
FQ211696.1
-
2
180
FQ211790.1
-
2
180
FQ211806.1
-
2
180
FQ211815.1
-
2
180
FQ211829.1
-
2
182
FQ211847.1
-
2
180
FQ211858.1
-
2
180
FQ211864.1
-
2
180
FQ211905.1
-
2
180
FQ211931.1
-
2
180
FQ211999.1
-
2
180
FQ212127.1
-
2
180
FQ212156.1
-
2
178
FQ212234.1
-
2
180
FQ212258.1
-
2
179
FQ212275.1
-
2
180
FQ212287.1
-
2
180
FQ212290.1
-
2
180
FQ212303.1
-
2
180
FQ212341.1
-
2
182
FQ212449.1
-
2
180
FQ213270.1
-
2
180
FQ213352.1
-
2
180
FQ213466.1
-
2
180
FQ213534.1
-
2
180
FQ213582.1
-
659
FQ213796.1
-
2
180
FQ214044.1
-
2
180
FQ214573.1
-
2
180
FQ216995.1
-
2
182
FQ217014.1
-
2
182
FQ217108.1
-
2
180
FQ217151.1
-
2
180
FQ217201.1
-
2
180
FQ217206.1
-
2
180
FQ217586.1
-
2
182
FQ217619.1
-
180
FQ217623.1
-
2
180
FQ217653.1
-
2
180
FQ217685.1
-
2
180
FQ217803.1
-
2
149
FQ218033.1
-
2
179
FQ218042.1
-
2
180
FQ218100.1
-
2
180
FQ218112.1
-
2
180
FQ218118.1
-
2
182
FQ218119.1
-
2
179
FQ218124.1
-
2
184
FQ218145.1
-
2
180
FQ218153.1
-
2
180
FQ218162.1
-
2
180
FQ218163.1
-
2
180
FQ218171.1
-
2
180
FQ218180.1
-
2
180
FQ218182.1
-
2
180
FQ218199.1
-
2
183
FQ218229.1
-
2
183
FQ218253.1
-
2
180
FQ218272.1
-
2
177
FQ218288.1
-
2
180
FQ218461.1
-
2
200
FQ218539.1
-
2
179
FQ218748.1
-
2
180
FQ218761.1
-
2
180
FQ218763.1
-
2
180
FQ218779.1
-
2
178
FQ219114.1
-
2
180
FQ219985.1
-
2
129
FQ220085.1
-
2
180
FQ220253.1
-
2
129
FQ221076.1
-
2
180
FQ221202.1
-
2
182
FQ221246.1
-
2
180
FQ221520.1
-
2
180
FQ221738.1
-
2
179
FQ221956.1
-
2
180
FQ221962.1
-
2
180
FQ222032.1
-
2
180
FQ222179.1
-
2
180
FQ222262.1
-
2
180
FQ222275.1
-
2
180
FQ222291.1
-
2
180
FQ222503.1
-
2
180
FQ222804.1
-
2
180
FQ222942.1
-
2
180
FQ222960.1
-
2
180
FQ223002.1
-
2
180
FQ223079.1
-
2
180
FQ223164.1
-
2
177
FQ223490.1
-
2
180
FQ223631.1
-
2
180
FQ223717.1
-
2
180
FQ223745.1
-
2
182
FQ223805.1
-
2
180
FQ223852.1
-
2
172
FQ223888.1
-
2
180
FQ223889.1
-
2
182
FQ223995.1
-
2
153
FQ224045.1
-
2
182
FQ224088.1
-
2
180
FQ224309.1
-
2
180
FQ224343.1
-
2
180
FQ224371.1
-
2
180
FQ224406.1
-
2
180
FQ224475.1
-
2
180
FQ224682.1
-
2
180
FQ224727.1
-
2
180
FQ224732.1
-
2
180
FQ225124.1
-
2
242
FQ225436.1
-
2
180
FQ225457.1
-
2
180
FQ225469.1
-
2
180
FQ225684.1
-
332
FQ225738.1
-
2
180
FQ225754.1
-
2
179
FQ225816.1
-
2
180
FQ225817.1
-
2
180
FQ226188.1
-
2
178
FQ226198.1
-
181
FQ226536.1
-
2
180
FQ226572.1
-
2
180
FQ226889.1
-
2
180
FQ227120.1
-
2
180
FQ227384.1
-
181
FQ227498.1
-
2
180
FQ227822.1
-
2
180
FQ227875.1
-
2
180
FQ228016.1
-
2
180
FQ228244.1
-
2
180
FQ228281.1
-
2
180
FQ228294.1
-
2
180
FQ228295.1
-
2
180
FQ228296.1
-
2
182
FQ228414.1
-
2
180
FQ228616.1
-
2
180
FQ228638.1
-
2
180
FQ229015.1
-
2
180
FQ229033.1
-
2
180
FQ229489.1
-
2
180
FQ230471.1
-
2
180
FQ230486.1
-
2
178
FQ230503.1
-
2
180
FQ230508.1
-
2
180
FQ230541.1
-
2
180
FQ230717.1
-
2
180
FQ230861.1
-
2
180
FQ230866.1
-
2
180
FQ230889.1
-
2
180
FQ231191.1
-
2
179
FQ231199.1
-
2
180
FQ231208.1
-
2
179
FQ231230.1
-
2
180
FQ231253.1
-
2
180
FQ231284.1
-
2
182
FQ231289.1
-
2
180
FQ232390.1
-
448
FQ232655.1
-
2
212
FQ233311.1
-
2
255
FQ234713.1
-
2
182
FQ234942.1
-
2
178
FQ235254.1
-
816
M17084.1
-
2
132
X16417.1
-
2
175
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: AB364477, AB020013.
Gene Ontology Categories:
Function(s)
Process(es)
glutathione metabolic process
regulation of eIF2 alpha phosphorylation by heme
oxygen transport
hemopoiesis
erythrocyte development
protein heterooligomerization
oxygen transporter activity
iron ion binding
oxygen binding
heme binding
hemoglobin alpha binding
hemoglobin beta binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-4vtwv:80/10.239.167.181:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline