Search Thermo Fisher Scientific
Gene Symbol: Ibsp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012587.2 | NP_036719.2 | 7 | 903 | ||
GenBank mRNA | AB001383.1 | - | 6 | 836 | ||
BC127506.1 | - | 7 | 899 | 8369751 | ||
DQ213013.1 | - | 6 | 830 | |||
DY313346.1 | - | 2 | 579 | |||
J04215.1 | - | 7 | 909 |