Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nefh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012607.2 | NP_036739.2 | 4 | 2931 | ||
XM_006251180.2 | XP_006251242.1 | 5 | 2867 | |||
XM_006251181.2 | XP_006251243.1 | 5 | 2849 | |||
XM_006251183.2 | XP_006251245.1 | 5 | 2777 | |||
XM_006251184.2 | XP_006251246.1 | 5 | 2759 | |||
XM_006251185.2 | XP_006251247.1 | 5 | 2705 | |||
XM_006251186.2 | XP_006251248.1 | 5 | 2705 | |||
XM_006251187.2 | XP_006251249.1 | 5 | 2633 | |||
XM_008770256.1 | XP_008768478.1 | 5 | 2921 | |||
XM_008770257.1 | XP_008768479.1 | 5 | 2579 | |||
GenBank mRNA | AF031879.1 | - | 2966 | |||
AY112897.1 | - | 1 | 1229 | |||
M21964.1 | - | 4 | 2072 | |||
M37227.1 | - | 4 | 2180 | |||
X13804.1 | - | 4 | 2180 |