Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde4d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113328.1 | NP_001106799.1 | 15 | 2195 | ||
NM_001113329.1 | NP_001106800.1 | 16 | 2110 | |||
NM_001113332.1 | NP_001106803.1 | 15 | 2047 | |||
NM_001113334.1 | NP_001106805.1 | 15 | 1884 | |||
NM_017032.1 | NP_058728.1 | 15 | 1802 | |||
XM_008760721.1 | XP_008758943.1 | 15 | 3203 | |||
XM_008760722.1 | XP_008758944.1 | 10 | 8906 | |||
XM_008760723.1 | XP_008758945.1 | 11 | 1546 | |||
GenBank mRNA | AF031373.1 | - | 15 | 2195 | ||
AF536974.1 | - | 10 | 1518 | |||
AF536979.1 | - | 16 | 2110 | |||
AY388961.1 | - | 15 | 1820 | |||
EF102484.1 | - | 15 | 2047 | |||
EF121818.1 | - | 15 | 1884 | |||
L27059.1 | - | 12 | 1521 | |||
L27060.1 | - | 10 | 1439 | |||
U09455.1 | - | 10 | 1688 | |||
U09456.1 | - | 11 | 1546 | |||
U09457.1 | - | 15 | 1802 |