Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pfkfb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033964.1 | NP_001029136.1 | 4 | 476 | ||
NM_001033965.1 | NP_001029137.1 | 4 | 476 | |||
NM_080477.3 | NP_536725.2 | 4 | 476 | |||
XM_006249706.2 | XP_006249768.1 | 4 | 436 | |||
XM_006249707.2 | XP_006249769.1 | 4 | 396 | |||
XM_006249708.1 | XP_006249770.1 | 4 | 476 | |||
XM_006249709.2 | XP_006249771.1 | 4 | 436 | |||
XM_006249710.1 | XP_006249772.1 | 3 | 361 | |||
XM_006249711.2 | XP_006249773.1 | 4 | 473 | |||
XM_008769425.1 | XP_008767647.1 | 3 | 275 | |||
GenBank mRNA | AB040530.1 | - | 4 | 476 | ||
AB040531.1 | - | 4 | 476 | |||
AB040532.1 | - | 4 | 476 | |||
AB040533.1 | - | 4 | 476 | |||
GQ422137.1 | - | 3 | 306 | |||
GQ422138.1 | - | 3 | 361 | |||
GQ438758.1 | - | 3 | 316 | |||
GQ438759.1 | - | 3 | 306 | |||
L27084.1 | - | 4 | 476 |