Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pfkfb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033964.1 | NP_001029136.1 | 11 | 1211 | ||
NM_001033965.1 | NP_001029137.1 | 11 | 1211 | |||
NM_080477.3 | NP_536725.2 | 11 | 1211 | |||
XM_006249706.2 | XP_006249768.1 | 11 | 1171 | |||
XM_006249707.2 | XP_006249769.1 | 11 | 1131 | |||
XM_006249708.1 | XP_006249770.1 | 11 | 1211 | |||
XM_006249709.2 | XP_006249771.1 | 11 | 1171 | |||
XM_006249710.1 | XP_006249772.1 | 10 | 1096 | |||
XM_006249711.2 | XP_006249773.1 | 11 | 1208 | |||
XM_008769425.1 | XP_008767647.1 | 10 | 1010 | |||
GenBank mRNA | AB040530.1 | - | 11 | 1211 | ||
AB040531.1 | - | 11 | 1211 | |||
AB040532.1 | - | 11 | 1211 | |||
AB040533.1 | - | 11 | 1211 | |||
GQ422137.1 | - | 9 | 916 | |||
GQ422138.1 | - | 10 | 1096 | |||
GQ438758.1 | - | 9 | 926 | |||
GQ438759.1 | - | 9 | 936 | |||
L27084.1 | - | 11 | 1211 |