Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012676.1 | NP_036808.1 | 9 | 455 | ||
XM_006249822.1 | XP_006249884.1 | 11 | 564 | |||
XM_006249823.1 | XP_006249885.1 | 11 | 565 | |||
XM_006249825.2 | XP_006249887.1 | 11 | 578 | |||
XM_006249828.1 | XP_006249890.1 | 10 | 552 | |||
XM_006249829.1 | XP_006249891.1 | 10 | 553 | |||
XM_006249833.2 | XP_006249895.1 | 10 | 560 | |||
XM_006249835.1 | XP_006249897.1 | 9 | 522 | |||
XM_006249836.1 | XP_006249898.1 | 9 | 523 | |||
XM_006249838.1 | XP_006249900.1 | 9 | 523 | |||
XM_006249840.2 | XP_006249902.1 | 12 | 594 | |||
XM_006249842.2 | XP_006249904.1 | 10 | 551 | |||
XM_008769535.1 | XP_008767757.1 | 11 | 565 | |||
XM_008769536.1 | XP_008767758.1 | 11 | 565 | |||
XM_008769537.1 | XP_008767759.1 | 10 | 553 | |||
XM_008769538.1 | XP_008767760.1 | 9 | 523 | |||
GenBank mRNA | BC161855.1 | - | 10 | 560 | 9035753 | |
EU295527.1 | - | 8 | 425 | |||
M26051.1 | - | 9 | 455 | |||
M26052.1 | - | 9 | 475 |