Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TNNT3 SIRNA Assays ›
Gene Symbol:
Tnnt3
Gene Name:
troponin T type 3 (skeletal, fast)
Gene Aliases:
Tnt
Chromosome Location:
Chr.1: 215666628 - 215683628 on Build Rnor_6.0
UniGene ID:
Rn.15488
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s128529
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Tnnt3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001270664.1
NP_001257593.1
12
642
NM_001270665.1
NP_001257594.1
14
675
NM_001270666.1
NP_001257595.1
14
675
NM_001270668.1
NP_001257597.1
12
642
NM_001270670.1
NP_001257599.1
13
657
NM_001270671.1
NP_001257600.1
13
657
NM_001270672.1
NP_001257601.1
12
648
NM_001270673.1
NP_001257602.1
13
660
NM_001270675.1
NP_001257604.1
12
648
NM_001270676.1
NP_001257605.1
13
660
NM_001270677.1
NP_001257606.1
11
630
NM_001270678.1
NP_001257607.1
13
663
NM_031532.2
NP_113720.1
13
663
XM_006230681.2
XP_006230743.1
12
605
XM_006230683.2
XP_006230745.1
11
593
XM_006230685.2
XP_006230747.1
10
575
XM_006230686.2
XP_006230748.1
10
575
XM_008760080.1
XP_008758302.1
15
709
XM_008760081.1
XP_008758303.1
15
709
XM_008760082.1
XP_008758304.1
14
659
XM_008760083.1
XP_008758305.1
14
656
XM_008760084.1
XP_008758306.1
13
682
XM_008760085.1
XP_008758307.1
13
647
XM_008760086.1
XP_008758308.1
12
629
GenBank mRNA
DQ062204.1
-
13
559
DQ273678.1
-
13
559
FQ214650.1
-
13
623
FQ214722.1
-
12
610
FQ214724.1
-
13
624
FQ214766.1
-
12
604
FQ214833.1
-
13
625
FQ215030.1
-
12
604
FQ215059.1
-
12
635
FQ215106.1
-
12
604
FQ215252.1
-
13
622
FQ215316.1
-
12
624
FQ215319.1
-
12
604
FQ215327.1
-
14
637
FQ215374.1
-
13
650
FQ215598.1
-
14
633
FQ215602.1
-
12
637
FQ215694.1
-
12
642
FQ215723.1
-
14
676
FQ215881.1
-
14
637
FQ215885.1
-
14
625
FQ215897.1
-
13
650
FQ215955.1
-
12
604
FQ216024.1
-
9
604
FQ216051.1
-
12
604
FQ216059.1
-
14
632
FQ216097.1
-
13
618
FQ216131.1
-
12
635
FQ216208.1
-
13
622
FQ216324.1
-
12
604
FQ216367.1
-
12
604
FQ216467.1
-
12
604
FQ216480.1
-
12
610
FQ216535.1
-
13
619
FQ216561.1
-
9
604
FQ216694.1
-
13
619
FQ216711.1
-
12
641
FQ216750.1
-
9
495
FQ216784.1
-
12
610
FQ216866.1
-
14
637
FQ216925.1
-
4
366
FQ217147.1
-
12
603
FQ217587.1
-
2
113
FQ217846.1
-
14
669
FQ217900.1
-
13
650
FQ223840.1
-
12
604
FQ223999.1
-
12
604
FQ224006.1
-
12
606
FQ224794.1
-
12
604
FQ224836.1
-
12
604
FQ224839.1
-
13
622
FQ224892.1
-
13
625
FQ224966.1
-
13
608
Back To Top
More Information
Panther Classification:
Molecular Function -
actin binding motor protein
Gene Ontology Categories:
Function(s)
Process(es)
skeletal muscle contraction
muscle contraction
regulation of muscle contraction
regulation of striated muscle contraction
metabolic process
regulation of ATPase activity
actin binding
tropomyosin binding
troponin C binding
calcium-dependent ATPase activity
troponin I binding
troponin T binding
calcium-dependent protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-46vz9:80/10.239.163.144:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0