Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ST3GAL3 SIRNA Assays ›
Gene Symbol:
ST3GAL3
Gene Name:
ST3 beta-galactoside alpha-2,3-sialyltransferase 3
Gene Aliases:
EIEE15, MRT12, SIAT6, ST3GALII, ST3GalIII, ST3N
Chromosome Location:
Chr.1: 43707497 - 43931165 on Build GRCh38
UniGene ID:
Hs.597915
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s12853
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ST3GAL3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001270459.1
NP_001257388.1
4
387
NM_001270460.1
NP_001257389.1
4
387
NM_001270461.1
NP_001257390.1
3
339
NM_001270462.1
NP_001257391.1
3
339
NM_001270463.1
NP_001257392.1
3
384
NM_001270464.1
NP_001257393.1
3
339
NM_001270465.1
NP_001257394.1
3
384
NM_001270466.1
NP_001257395.1
3
339
NM_006279.3
NP_006270.1
4
387
NM_174963.3
NP_777623.2
4
432
NM_174964.2
NP_777624.1
4
432
NM_174965.2
NP_777625.1
4
432
NM_174966.2
NP_777626.1
4
387
NM_174967.2
NP_777627.1
4
387
NM_174968.3
NP_777628.2
4
387
NM_174969.2
NP_777629.1
3
339
NM_174970.2
NP_777630.1
3
339
NM_174971.3
NP_777631.2
3
339
NR_073016.1
4
387
NR_073018.1
4
387
NR_073020.1
3
339
NR_073021.1
4
387
NR_073023.1
4
387
XM_006710825.3
XP_006710888.1
4
423
XM_006710827.3
XP_006710890.1
3
341
XM_006710828.3
XP_006710891.1
4
389
XM_011541973.2
XP_011540275.1
4
434
XM_011541974.2
XP_011540276.1
4
389
XM_011541976.2
XP_011540278.1
3
341
XM_011541977.2
XP_011540279.1
4
434
XM_011541981.2
XP_011540283.1
4
434
XM_011541983.2
XP_011540285.1
5
698
XM_011541984.2
XP_011540286.1
4
434
XM_011541985.2
XP_011540287.1
4
434
XM_011541986.2
XP_011540288.1
4
434
XM_011541987.2
XP_011540289.1
4
389
XM_017002109.1
XP_016857598.1
4
497
XM_017002110.1
XP_016857599.1
4
434
XM_017002111.1
XP_016857600.1
4
490
XM_017002112.1
XP_016857601.1
4
486
XM_017002113.1
XP_016857602.1
4
467
XM_017002114.1
XP_016857603.1
4
490
XM_017002115.1
XP_016857604.1
3
341
XM_017002116.1
XP_016857605.1
4
486
XM_017002117.1
XP_016857606.1
4
389
XM_017002118.1
XP_016857607.1
4
434
XM_017002119.1
XP_016857608.1
4
387
XM_017002120.1
XP_016857609.1
4
386
XM_017002121.1
XP_016857610.1
2
179
XM_017002122.1
XP_016857611.1
4
490
GenBank mRNA
AF425851.1
-
3
241
AF425852.1
-
3
241
AF425853.1
-
3
241
AF425854.1
-
3
196
AF425855.1
-
3
196
AF425856.1
-
3
196
AF425857.1
-
3
196
AF425858.1
-
3
196
AF425859.1
-
2
148
AF425860.1
-
2
148
AF425861.1
-
2
148
AF425862.1
-
3
196
AF425863.1
-
2
193
AF425864.1
-
3
196
AF425865.1
-
3
196
AF425866.1
-
3
196
AF425867.1
-
3
196
AK301829.1
-
4
373
AK310265.1
-
5
665
AY051143.1
-
3
324
AY167992.1
-
3
196
AY167993.1
-
2
148
AY167994.1
-
2
193
AY167995.1
-
2
148
AY167996.1
-
2
148
AY167997.1
-
2
148
AY167998.1
-
2
148
BC050380.1
-
4
371
5769575
L23768.1
-
3
196
Back To Top
More Information
Panther Classification:
Molecular Function -
transferase
metabolite interconversion enzyme
Gene Ontology Categories:
Function(s)
Process(es)
ganglioside biosynthetic process
oligosaccharide metabolic process
O-glycan processing
keratan sulfate biosynthetic process
protein N-linked glycosylation via asparagine
sialylation
beta-galactoside (CMP) alpha-2,3-sialyltransferase activity
N-acetyllactosaminide alpha-2,3-sialyltransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-qskjf:80/10.239.165.3:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0