Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TPM1 SIRNA Assays ›
Gene Symbol:
Tpm1
Gene Name:
tropomyosin 1, alpha
Gene Aliases:
Alpha-tm, Tma2, Tmsa
Chromosome Location:
Chr.8: 72814737 - 72841496 on Build Rnor_6.0
UniGene ID:
Rn.87540
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s128551
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Tpm1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001034068.1
NP_001029240.1
5
621
NM_001034069.1
NP_001029241.1
5
621
NM_001034070.1
NP_001029242.1
5
621
NM_001034071.1
NP_001029243.1
5
621
NM_001034072.1
NP_001029244.1
5
621
NM_001034073.1
NP_001029245.1
4
558
NM_001034074.1
NP_001029246.1
4
558
NM_001034075.1
NP_001029247.1
4
558
NM_001301336.1
NP_001288265.1
5
621
NM_001301342.1
NP_001288271.1
5
621
NM_001301736.1
NP_001288665.1
4
558
NM_019131.2
NP_062004.1
4
558
XM_006243386.2
XP_006243448.1
5
613
XM_006243388.2
XP_006243450.1
5
613
XM_006243389.2
XP_006243451.1
5
613
XM_006243390.2
XP_006243452.1
5
613
XM_006243393.1
XP_006243455.1
4
556
XM_006243394.1
4
574
XM_006243395.2
XP_006243457.1
4
565
XM_008766207.1
XP_008764429.1
5
613
XM_008766208.1
XP_008764430.1
4
556
GenBank mRNA
AF372215.1
-
4
407
AF372216.1
-
5
520
AF372217.1
-
5
526
BC078780.1
-
4
525
7106337
CB585296.1
-
4
561
CF108680.1
-
5
569
CK483405.1
-
5
575
FQ074026.1
-
5
679
FQ214619.1
-
5
596
FQ214809.1
-
5
590
FQ214835.1
-
5
590
FQ214864.1
-
5
594
FQ214868.1
-
5
590
FQ214869.1
-
5
596
FQ214894.1
-
5
596
FQ214902.1
-
5
590
FQ214911.1
-
5
592
FQ214944.1
-
5
596
FQ214946.1
-
5
596
FQ214991.1
-
5
590
FQ215107.1
-
5
590
FQ215186.1
-
5
596
FQ215188.1
-
5
590
FQ215206.1
-
5
625
FQ215226.1
-
5
596
FQ215459.1
-
5
592
FQ215512.1
-
5
596
FQ215522.1
-
5
667
FQ215524.1
-
5
592
FQ215538.1
-
5
596
FQ215551.1
-
5
591
FQ215698.1
-
5
590
FQ215703.1
-
5
590
FQ215724.1
-
5
677
FQ215746.1
-
5
592
FQ215761.1
-
5
590
FQ215768.1
-
5
590
FQ215803.1
-
5
596
FQ215808.1
-
5
592
FQ215815.1
-
5
590
FQ215919.1
-
5
598
FQ215944.1
-
5
597
FQ215969.1
-
5
590
FQ215995.1
-
5
592
FQ216014.1
-
5
674
FQ216018.1
-
5
596
FQ216052.1
-
5
590
FQ216105.1
-
5
591
FQ216118.1
-
5
674
FQ216145.1
-
5
596
FQ216160.1
-
5
589
FQ216319.1
-
5
592
FQ216340.1
-
5
596
FQ216368.1
-
5
592
FQ216400.1
-
5
592
FQ216494.1
-
5
590
FQ216552.1
-
5
596
FQ216557.1
-
5
596
FQ216569.1
-
685
FQ216615.1
-
5
505
FQ216637.1
-
5
592
FQ216683.1
-
5
592
FQ216697.1
-
5
592
FQ216747.1
-
5
592
FQ217468.1
-
4
341
FQ217503.1
-
5
596
FQ217616.1
-
3
158
FQ217637.1
-
5
596
FQ217808.1
-
5
592
FQ223706.1
-
5
592
FQ223993.1
-
5
594
FQ224036.1
-
5
507
FQ224207.1
-
5
590
FQ224838.1
-
5
596
FQ224943.1
-
5
594
FQ224961.1
-
5
592
M34134.1
-
4
543
M34135.1
-
5
555
M34136.1
-
3
273
M60666.1
-
5
568
M60667.1
-
5
568
M60668.1
-
4
505
M60669.1
-
4
505
Back To Top
More Information
Panther Classification:
Molecular Function -
actin binding motor protein
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
positive regulation of heart rate by epinephrine
muscle contraction
muscle filament sliding
negative regulation of cell migration
ruffle organization
positive regulation of ATPase activity
cellular response to reactive oxygen species
wound healing
regulation of ATPase activity
sarcomere organization
positive regulation of cell adhesion
positive regulation of stress fiber assembly
actin filament capping
ventricular cardiac muscle tissue morphogenesis
cardiac muscle contraction
actin binding
cytoskeletal protein binding
protein homodimerization activity
protein N-terminus binding
actin filament binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-l9dfq:80/10.239.161.188:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0