Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TTR SIRNA Assays ›
Gene Symbol:
Ttr
Gene Name:
transthyretin
Gene Aliases:
Lr1, Tbpa
Chromosome Location:
Chr.18: 15532963 - 15542180 on Build Rnor_6.0
UniGene ID:
Rn.1404
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s128562
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Ttr
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_012681.2
NP_036813.2
2
198
XM_006254457.1
XP_006254519.1
3
255
GenBank mRNA
AF479660.1
-
2
133
BC086946.1
-
2
91
7322736
FQ209711.1
-
2
113
FQ209724.1
-
2
116
FQ209731.1
-
2
116
FQ209737.1
-
2
116
FQ209743.1
-
2
116
FQ209798.1
-
2
116
FQ209827.1
-
2
115
FQ209853.1
-
2
116
FQ209868.1
-
2
116
FQ209884.1
-
2
116
FQ209896.1
-
2
109
FQ209911.1
-
2
116
FQ209919.1
-
2
115
FQ209931.1
-
2
113
FQ209949.1
-
2
116
FQ209960.1
-
2
115
FQ209974.1
-
2
116
FQ209986.1
-
2
116
FQ209992.1
-
2
113
FQ210012.1
-
2
115
FQ210053.1
-
2
116
FQ210063.1
-
2
116
FQ210074.1
-
2
111
FQ210079.1
-
2
116
FQ210088.1
-
2
116
FQ210092.1
-
2
116
FQ210104.1
-
2
115
FQ210128.1
-
2
116
FQ210130.1
-
2
116
FQ210144.1
-
2
116
FQ210148.1
-
2
116
FQ210149.1
-
2
116
FQ210166.1
-
2
116
FQ210168.1
-
2
116
FQ210188.1
-
2
116
FQ210192.1
-
2
111
FQ210193.1
-
2
116
FQ210202.1
-
2
116
FQ210212.1
-
2
116
FQ210220.1
-
2
116
FQ210221.1
-
2
116
FQ210257.1
-
2
116
FQ210271.1
-
2
116
FQ210283.1
-
2
113
FQ210289.1
-
2
111
FQ210300.1
-
2
116
FQ210301.1
-
2
116
FQ210303.1
-
2
116
FQ210314.1
-
2
116
FQ210319.1
-
2
116
FQ210324.1
-
2
114
FQ210363.1
-
2
116
FQ210365.1
-
2
115
FQ210370.1
-
2
116
FQ210380.1
-
2
116
FQ210383.1
-
2
113
FQ210386.1
-
566
FQ210389.1
-
2
116
FQ210423.1
-
2
115
FQ210429.1
-
2
116
FQ210431.1
-
2
116
FQ210434.1
-
2
116
FQ210451.1
-
2
116
FQ210464.1
-
2
111
FQ210465.1
-
2
116
FQ210502.1
-
2
116
FQ210545.1
-
2
116
FQ210547.1
-
2
116
FQ210548.1
-
2
113
FQ210552.1
-
2
111
FQ210579.1
-
2
115
FQ210580.1
-
2
113
FQ210584.1
-
2
116
FQ210588.1
-
2
116
FQ210614.1
-
2
116
FQ210642.1
-
2
116
FQ210659.1
-
2
115
FQ210663.1
-
2
113
FQ210667.1
-
2
116
FQ210680.1
-
2
111
FQ210689.1
-
2
116
FQ210691.1
-
2
116
FQ210711.1
-
2
113
FQ210747.1
-
2
114
FQ210750.1
-
2
115
FQ210759.1
-
2
116
FQ210777.1
-
2
113
FQ210778.1
-
2
116
FQ210779.1
-
2
116
FQ210786.1
-
2
116
FQ210844.1
-
2
116
FQ210853.1
-
2
113
FQ210860.1
-
2
116
FQ210869.1
-
2
116
FQ210887.1
-
2
116
FQ210913.1
-
2
116
FQ210915.1
-
2
116
FQ210971.1
-
2
116
FQ210974.1
-
2
116
FQ210975.1
-
2
116
FQ210993.1
-
2
116
FQ211000.1
-
2
116
FQ211011.1
-
2
116
FQ211023.1
-
2
116
FQ211028.1
-
2
113
FQ211048.1
-
2
116
FQ211066.1
-
2
116
FQ211087.1
-
2
116
FQ211100.1
-
2
116
FQ211107.1
-
2
116
FQ211134.1
-
2
115
FQ211143.1
-
2
116
FQ211145.1
-
2
116
FQ211149.1
-
2
116
FQ211173.1
-
2
116
FQ211184.1
-
2
122
FQ211191.1
-
2
116
FQ211204.1
-
2
116
FQ211212.1
-
2
116
FQ211228.1
-
2
116
FQ211233.1
-
2
118
FQ211237.1
-
2
113
FQ211240.1
-
2
116
FQ211260.1
-
2
116
FQ211262.1
-
2
114
FQ211291.1
-
2
113
FQ211302.1
-
2
116
FQ211335.1
-
2
116
FQ211341.1
-
2
116
FQ211348.1
-
2
113
FQ211403.1
-
2
116
FQ211448.1
-
2
116
FQ211620.1
-
2
116
FQ212302.1
-
2
116
FQ213410.1
-
2
201
FQ217460.1
-
2
116
FQ218087.1
-
2
116
FQ218127.1
-
2
117
FQ218128.1
-
2
112
FQ218131.1
-
2
116
FQ218133.1
-
2
116
FQ218134.1
-
2
116
FQ218142.1
-
2
111
FQ218150.1
-
2
116
FQ218178.1
-
2
116
FQ218187.1
-
2
116
FQ218296.1
-
2
116
FQ218301.1
-
2
116
FQ218306.1
-
2
116
FQ218313.1
-
2
116
FQ218333.1
-
2
111
FQ218513.1
-
2
111
FQ218589.1
-
2
116
FQ218636.1
-
2
116
FQ218738.1
-
2
116
FQ218740.1
-
2
116
FQ218747.1
-
2
116
FQ218749.1
-
2
116
FQ218758.1
-
2
41
FQ218786.1
-
2
116
FQ218849.1
-
2
116
FQ218866.1
-
2
116
FQ218996.1
-
2
116
FQ219153.1
-
2
113
FQ219387.1
-
2
116
FQ219428.1
-
2
116
FQ223520.1
-
2
111
FQ223553.1
-
2
116
FQ224434.1
-
2
116
K03252.1
-
2
96
X14876.1
-
2
89
Back To Top
More Information
Panther Classification:
Molecular Function -
hydrolase
Gene Ontology Categories:
Function(s)
Process(es)
thyroid hormone metabolic process
retinol metabolic process
thyroid hormone transport
hormone activity
hormone binding
identical protein binding
protein heterodimerization activity
thyroid hormone binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6d74b6fc5b-sshrn:80/10.239.165.44:80.
git-commit: 0683b6a9b3dadc77a268b704485bdfd9f1fd09b0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.0-2024.10.41-1.0