Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031534.2 | NP_113722.2 | 9 | 1229 | ||
XM_006234620.1 | XP_006234682.1 | 8 | 1571 | |||
GenBank mRNA | CO382418.1 | - | 1 | 97 | ||
X69716.1 | - | 9 | 1229 |