Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033670.1 | NP_001028842.1 | 2 | 263 | ||
NM_001033671.1 | NP_001028843.1 | 1 | 197 | |||
NM_001033672.1 | NP_001028844.1 | 2 | 420 | |||
NM_031535.2 | NP_113723.2 | 2 | 248 | |||
XM_006235264.2 | XP_006235326.1 | 2 | 262 | |||
XM_006235265.2 | XP_006235327.1 | 2 | 1075 | |||
GenBank mRNA | AF136230.1 | - | 1 | 85 | ||
AF279286.1 | - | 1 | 85 | |||
AY141038.1 | - | 2 | 420 | |||
BC094213.1 | - | 2 | 263 | 7314572 | ||
S76513.1 | - | 1 | 104 | |||
S78284.1 | - | 1 | 104 | |||
U34963.1 | - | 1 | 104 | |||
U72349.1 | - | 2 | 248 | |||
U72350.1 | - | 1 | 156 |