Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cyp2c12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031572.1 | NP_113760.1 | 4 | 595 | ||
GenBank mRNA | BC089790.1 | - | 4 | 603 | 7371667 | |
FQ209627.1 | - | 1 | 34 | |||
FQ219038.1 | - | 2 | 276 | |||
FQ219491.1 | - | 3 | 470 | |||
FQ219720.1 | - | 1 | 65 | |||
J03786.1 | - | 4 | 595 |