Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plcb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024353.1 | NP_077329.1 | 26 | 2494 | ||
XM_006235083.2 | XP_006235145.1 | 27 | 2564 | |||
XM_006235085.2 | XP_006235147.1 | 27 | 2425 | |||
XM_006235088.2 | XP_006235150.1 | 26 | 4678 | |||
XM_006235089.2 | XP_006235151.1 | 27 | 5251 | |||
XM_006235094.2 | XP_006235156.1 | 26 | 2528 | |||
XM_008762248.1 | XP_008760470.1 | 27 | 4797 | |||
XM_008762249.1 | XP_008760471.1 | 26 | 4761 | |||
XM_008762250.1 | XP_008760472.1 | 27 | 4797 | |||
XM_008762251.1 | XP_008760473.1 | 26 | 4761 | |||
GenBank mRNA | AF027571.1 | - | 9 | 883 | ||
L15556.1 | - | 26 | 2494 |