Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plcb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024353.1 | NP_077329.1 | 21 | 1925 | ||
XM_006235083.2 | XP_006235145.1 | 22 | 1995 | |||
XM_006235085.2 | XP_006235147.1 | 22 | 1856 | |||
XM_006235088.2 | XP_006235150.1 | 21 | 4109 | |||
XM_006235089.2 | XP_006235151.1 | 22 | 4682 | |||
XM_006235094.2 | XP_006235156.1 | 21 | 1959 | |||
XM_008762248.1 | XP_008760470.1 | 22 | 4228 | |||
XM_008762249.1 | XP_008760471.1 | 21 | 4192 | |||
XM_008762250.1 | XP_008760472.1 | 22 | 4228 | |||
XM_008762251.1 | XP_008760473.1 | 21 | 4192 | |||
GenBank mRNA | AF027571.1 | - | 4 | 314 | ||
AF031370.1 | - | 19 | 1692 | |||
L15556.1 | - | 21 | 1925 | |||
L18962.1 | - | 21 | 1825 | |||
U57836.1 | - | 21 | 1825 |