Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019630.3 | NP_062604.1 | 6 | 560 | ||
XM_006247177.2 | XP_006247239.1 | 5 | 782 | |||
XM_008767947.1 | XP_008766169.1 | 6 | 602 | |||
GenBank mRNA | D11439.1 | - | 5 | 532 | ||
L08831.1 | - | 6 | 560 | |||
M92916.1 | - | 5 | 2732 | |||
X66724.1 | - | 6 | 621 | |||
Z19564.1 | - | 6 | 560 |