Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apoa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012738.1 | NP_036870.1 | 3 | 189 | ||
GenBank mRNA | BC089820.1 | - | 2 | 173 | 7321674 | |
FQ209434.1 | - | 3 | 190 | |||
FQ209676.1 | - | 3 | 194 | |||
FQ209742.1 | - | 3 | 194 | |||
FQ209761.1 | - | 3 | 190 | |||
FQ209776.1 | - | 3 | 194 | |||
FQ209818.1 | - | 3 | 195 | |||
FQ209900.1 | - | 3 | 190 | |||
FQ209928.1 | - | 3 | 194 | |||
FQ209958.1 | - | 3 | 194 | |||
FQ210010.1 | - | 3 | 194 | |||
FQ210126.1 | - | 3 | 191 | |||
FQ210161.1 | - | 3 | 217 | |||
FQ210354.1 | - | 3 | 194 | |||
FQ210367.1 | - | 3 | 190 | |||
FQ210371.1 | - | 3 | 190 | |||
FQ210443.1 | - | 3 | 191 | |||
FQ210470.1 | - | 3 | 190 | |||
FQ210544.1 | - | 3 | 192 | |||
FQ210653.1 | - | 3 | 192 | |||
FQ210722.1 | - | 3 | 194 | |||
FQ210743.1 | - | 3 | 297 | |||
FQ210863.1 | - | 3 | 194 | |||
FQ210880.1 | - | 3 | 190 | |||
FQ210914.1 | - | 3 | 190 | |||
FQ210920.1 | - | 3 | 190 | |||
FQ211297.1 | - | 3 | 192 | |||
FQ211535.1 | - | 3 | 197 | |||
FQ211571.1 | - | 3 | 190 | |||
FQ218078.1 | - | 3 | 190 | |||
FQ218079.1 | - | 3 | 194 | |||
FQ218325.1 | - | 3 | 194 | |||
FQ218519.1 | - | 3 | 190 | |||
FQ218525.1 | - | 3 | 190 | |||
FQ218615.1 | - | 3 | 189 | |||
FQ218659.1 | - | 3 | 193 | |||
FQ218663.1 | - | 3 | 190 | |||
FQ218837.1 | - | 3 | 196 | |||
FQ219021.1 | - | 3 | 194 | |||
FQ219052.1 | - | 3 | 196 | |||
FQ219074.1 | - | 3 | 190 | |||
FQ219479.1 | - | 3 | 282 | |||
FQ219515.1 | - | 3 | 194 | |||
FQ219577.1 | - | 3 | 190 | |||
FQ219728.1 | - | 3 | 189 | |||
FQ224655.1 | - | 3 | 190 | |||
FQ229884.1 | - | 3 | 229 | |||
M00001.1 | - | 3 | 189 | |||
X00558.1 | - | 2 | 178 |