Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ampd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031544.1 | NP_113732.1 | 9 | 1421 | ||
XM_006230010.2 | XP_006230072.1 | 9 | 2189 | |||
XM_006230011.2 | XP_006230073.1 | 9 | 1554 | |||
XM_006230012.2 | XP_006230074.1 | 9 | 2024 | |||
XM_006230013.2 | XP_006230075.1 | 9 | 2036 | |||
XM_006230014.2 | XP_006230076.1 | 9 | 1940 | |||
XM_006230015.2 | XP_006230077.1 | 9 | 2168 | |||
XM_008759688.1 | XP_008757910.1 | 9 | 1431 | |||
GenBank mRNA | U90888.1 | - | 9 | 1421 |