Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030992.1 | NP_112254.1 | 4 | 491 | ||
XM_006232205.2 | XP_006232267.1 | 4 | 1266 | |||
XM_006232206.2 | XP_006232268.1 | 5 | 919 | |||
XM_008760891.1 | XP_008759113.1 | 5 | 919 | |||
GenBank mRNA | AB000778.1 | - | 4 | 506 | ||
AB000779.1 | - | 4 | 491 | |||
AB003170.1 | - | 4 | 383 | |||
AB003171.1 | - | 4 | 383 | |||
AF017251.1 | - | 3 | 335 | |||
U69550.1 | - | 4 | 491 | |||
U88986.1 | - | 4 | 403 |