Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012744.2 | NP_036876.2 | 5 | 617 | ||
XM_006230689.2 | XP_006230751.1 | 6 | 848 | |||
XM_006230690.2 | XP_006230752.1 | 5 | 963 | |||
XM_006230691.2 | XP_006230753.1 | 6 | 670 | |||
XM_006230692.2 | XP_006230754.1 | 5 | 640 | |||
XM_006230693.2 | XP_006230755.1 | 6 | 772 | |||
XM_006230694.2 | XP_006230756.1 | 5 | 639 | |||
XM_008760087.1 | XP_008758309.1 | 5 | 745 | |||
GenBank mRNA | BC085680.1 | - | 5 | 594 | 7109369 | |
U32314.1 | - | 5 | 585 | |||
U36585.1 | - | 5 | 617 |