Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031546.1 | NP_113734.1 | 4 | 590 | ||
GenBank mRNA | AB037934.1 | - | 4 | 590 | ||
BC078794.1 | - | 4 | 585 | 7126233 | ||
D38467.1 | - | 4 | 584 | |||
FQ219778.1 | - | 4 | 593 | |||
X69021.1 | - | 4 | 579 |