Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inhbb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080771.1 | NP_542949.1 | 2 | 652 | ||
GenBank mRNA | AF478684.1 | - | 1 | 183 |