Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013026.2 | NP_037158.1 | 3, 4 | 846 | ||
GenBank mRNA | BC061834.1 | - | 3, 4 | 846 | 5600086 | |
M81785.1 | - | 3, 4 | 861 | |||
S61865.1 | - | 3, 4 | 861 | |||
X60651.1 | - | 2, 3 | 536 |