Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Add3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164103.1 | NP_001157575.1 | 10 | 1541 | ||
NM_031552.2 | NP_113740.2 | 10 | 1541 | |||
XM_006231599.2 | XP_006231661.1 | 10 | 1394 | |||
XM_006231600.2 | XP_006231662.1 | 11 | 1641 | |||
XM_006231601.2 | XP_006231663.1 | 10 | 2240 | |||
XM_006231602.2 | XP_006231664.1 | 10 | 2242 | |||
XM_008760510.1 | XP_008758732.1 | 10 | 1384 | |||
XM_008760511.1 | XP_008758733.1 | 10 | 2239 | |||
GenBank mRNA | CO402732.1 | - | 1 | 27 | ||
U35775.1 | - | 1458 |