Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC2A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006516.2 | NP_006507.2 | 9 | 1617 | ||
GenBank mRNA | AB208987.1 | - | 9 | 1337 | ||
AF070544.1 | - | 6 | 645 | |||
AK122999.1 | - | 7 | 877 | |||
AK292791.1 | - | 9 | 1303 | |||
AK293306.1 | - | 9 | 1300 | |||
AK296736.1 | - | 10 | 1135 | |||
AK312403.1 | - | 9 | 1309 | |||
AY034633.1 | - | 3 | 213 | |||
BC118590.1 | - | 9 | 1238 | 40085221 | ||
BG682043.1 | - | 5 | 460 | |||
K03195.1 | - | 9 | 1271 |