Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031851.2 | NP_114039.1 | 7 | 983 | ||
GenBank mRNA | BC072518.1 | - | 7 | 983 | 7105633 | |
BC097304.1 | - | 7 | 1401 | 7441791 | ||
FQ213801.1 | - | 7 | 998 | |||
FQ215559.1 | - | 7 | 1000 | |||
FQ229103.1 | - | 7 | 985 | |||
FQ229278.1 | - | 7 | 1018 | |||
FQ231383.1 | - | 7 | 976 | |||
FQ232197.1 | - | 7 | 976 | |||
M61219.1 | - | 6 | 935 |