Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012924.2 | NP_037056.2 | 12 | 1600 | ||
XM_006234627.2 | XP_006234689.1 | 9 | 1373 | |||
XM_006234629.2 | XP_006234691.1 | 7 | 1139 | |||
XM_008762055.1 | XP_008760277.1 | 9 | 1372 | |||
XM_008762059.1 | XP_008760281.1 | 12 | 1737 | |||
GenBank mRNA | AF014365.1 | - | 7 | 806 | ||
M61874.1 | - | 9 | 1228 | |||
U96138.1 | - | 12 | 1600 |