Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctsh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012939.1 | NP_037071.1 | 8 | 611 | ||
XM_008766440.1 | XP_008764662.1 | 8 | 662 | |||
GenBank mRNA | BC085352.1 | - | 8 | 626 | 7115929 | |
FQ210730.1 | - | 8 | 621 | |||
FQ213183.1 | - | 8 | 621 | |||
FQ219177.1 | - | 8 | 621 | |||
FQ219285.1 | - | 8 | 621 | |||
FQ219548.1 | - | 8 | 621 | |||
FQ219611.1 | - | 8 | 639 | |||
FQ220588.1 | - | 5 | 390 | |||
FQ223707.1 | - | 8 | 617 | |||
FQ228686.1 | - | 8 | 681 | |||
FQ228906.1 | - | 8 | 628 | |||
FQ228938.1 | - | 8 | 618 | |||
FQ228956.1 | - | 8 | 648 | |||
M38135.1 | - | 8 | 605 | |||
Y00708.1 | - | 8 | 611 |