Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Drd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012944.1 | NP_037076.1 | 4 | 1238 | ||
XM_006230521.1 | XP_006230583.1 | 4 | 1208 | |||
XM_006230522.1 | XP_006230584.1 | 5 | 1124 | |||
GenBank mRNA | M84009.1 | - | 4 | 1238 |
Set Membership: |
G-protein coupled receptor |