Gene Symbol: Nrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012993.2 | NP_037125.1 | 22, 23 | 2704 | ||
XM_006238499.2 | XP_006238561.1 | 24, 25 | 3042 | |||
XM_006238500.2 | XP_006238562.1 | 23, 24 | 2906 | |||
GenBank mRNA | FQ218095.1 | - | 22, 23 | 2827 | ||
L27124.1 | - | 22, 23 | 2705 | |||
X93208.1 | - | 24, 25 | 2909 |