Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC7A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003045.4 | NP_003036.1 | 4 | 897 | ||
XM_005266507.3 | XP_005266564.1 | 4 | 692 | |||
XM_017020713.1 | XP_016876202.1 | 4 | 897 | |||
XM_017020714.1 | XP_016876203.1 | 4 | 836 | |||
XM_017020715.1 | XP_016876204.1 | 4 | 718 | |||
XM_017020716.1 | XP_016876205.1 | 4 | 908 | |||
GenBank mRNA | AB105377.1 | - | 4 | 784 | ||
AF078107.1 | - | 3 | 630 | |||
AK312818.1 | - | 4 | 671 | |||
BC063303.1 | - | 4 | 694 | 30340706 | ||
BC069358.1 | - | 3 | 579 | 7262451 | ||
BC115407.1 | - | 3 | 579 | 40076678 | ||
X57303.1 | - | 4 | 660 | |||
X59155.1 | - | 4 | 657 |