Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC8A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015063.2 | NP_055878.1 | 5 | 2053 | ||
XM_005259172.1 | XP_005259229.1 | 5 | 1862 | |||
XM_017027159.1 | XP_016882648.1 | 2 | 201 | |||
GenBank mRNA | AB029010.1 | - | 5 | 1868 | ||
AK293457.1 | - | 5 | 794 | |||
AK293836.1 | - | 3 | 266 | |||
AK293860.1 | - | 4 | 1186 | |||
AK302552.1 | - | 5 | 2053 | |||
AL512764.1 | - | 4 | 1163 |