Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: Stxbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013038.3 | NP_037170.1 | 6 | 507 | ||
XM_006233939.2 | XP_006234001.1 | 6 | 577 | |||
GenBank mRNA | BC088850.1 | - | 6 | 507 | 7315868 | |
L26264.1 | - | 6 | 486 | |||
U06069.1 | - | 6 | 418 | |||
U21116.1 | - | 6 | 401 |