- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: Ppp1cb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013065.2 | NP_037197.1 | 6, 7 | 1019 | ||
XM_008764505.1 | XP_008762727.1 | 7, 8 | 957 | |||
GenBank mRNA | BC062033.1 | - | 6, 7 | 1019 | 5623630 | |
D90164.1 | - | 6, 7 | 799 | |||
FQ212294.1 | - | 7, 8 | 957 | |||
M58434.1 | - | 1, 2 | 131 | |||
S78218.1 | - | 6, 7 | 786 |