Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hcrtr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013074.1 | NP_037206.1 | 3, 4 | 711 | ||
XM_008766215.1 | XP_008764437.1 | 3, 4 | 1071 | |||
GenBank mRNA | AF041246.1 | - | 3, 4 | 711 |
Set Membership: |
G-protein coupled receptor |