Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il1r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013123.3 | NP_037255.3 | 11 | 1366 | ||
XM_006244746.2 | XP_006244808.1 | 12 | 3475 | |||
XM_006244747.2 | XP_006244809.1 | 12 | 1732 | |||
XM_006244748.2 | XP_006244810.1 | 11 | 1636 | |||
XM_006244749.2 | XP_006244811.1 | 13 | 1555 | |||
XM_006244750.2 | XP_006244812.1 | 13 | 3547 | |||
XM_006244751.2 | XP_006244813.1 | 12 | 1466 | |||
XM_006244752.2 | XP_006244814.1 | 12 | 1708 | |||
XM_006244753.2 | XP_006244815.1 | 12 | 2827 | |||
XM_006244754.2 | XP_006244816.1 | 11 | 3379 | |||
XM_008766993.1 | XP_008765215.1 | 12 | 1483 | |||
GenBank mRNA | AB112436.1 | - | 12 | 1356 | ||
AB112437.1 | - | 11 | 1366 | |||
AB112438.1 | - | 12 | 1438 | |||
M95578.1 | - | 12 | 1687 |