Gene Symbol: Cald1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013146.2 | NP_037278.1 | 6, 7 | 1302 | ||
XM_006236260.2 | XP_006236322.1 | 6, 7 | 2852 | |||
XM_006236261.2 | XP_006236323.1 | 7, 8 | 1373 | |||
XM_006236262.2 | XP_006236324.1 | 7, 8 | 1526 | |||
XM_008762763.1 | XP_008760985.1 | 8, 9 | 1454 |