Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019122.1 | NP_061995.1 | 4 | 1130 | ||
XM_006228978.2 | XP_006229040.1 | 5 | 1588 | |||
XM_006228979.2 | XP_006229041.1 | 5 | 1593 | |||
XM_008759324.1 | XP_008757546.1 | 6 | 1318 | |||
XM_008759325.1 | XP_008757547.1 | 5 | 1222 | |||
GenBank mRNA | AF375464.1 | - | 3 | 1028 | ||
D28512.1 | - | 4 | 1130 | |||
EU441216.1 | - | 3 | 1033 |