Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tac3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019162.2 | NP_062035.1 | 5 | 387 | ||
GenBank mRNA | FQ224503.1 | - | 2 | 129 | ||
M16410.1 | - | 5 | 387 |