Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ak4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017135.3 | NP_058831.1 | 2 | 169 | ||
XM_006238445.2 | XP_006238507.1 | 3 | 474 | |||
GenBank mRNA | BC087024.1 | - | 2 | 221 | 7107348 | |
D87809.1 | - | 2 | 169 |