Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RPSA SIRNA Assays ›
Gene Symbol:
Rpsa
Gene Name:
ribosomal protein SA
Gene Aliases:
Lamr1
Chromosome Location:
Chr.8: 128806056 - 128809987 on Build Rnor_6.0
UniGene ID:
Rn.121972
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s130985
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Rpsa
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_017138.2
NP_058834.1
4
431
XM_008766672.1
XP_008764894.1
4
504
GenBank mRNA
BC060578.1
-
4
449
6919235
D25224.1
-
4
432
FJ386454.1
-
3
412
FQ209463.1
-
4
445
FQ209469.1
-
4
445
FQ209490.1
-
4
446
FQ209667.1
-
4
445
FQ209729.1
-
4
445
FQ211189.1
-
4
445
FQ212660.1
-
4
445
FQ212893.1
-
4
445
FQ213275.1
-
4
445
FQ213334.1
-
4
445
FQ213623.1
-
4
448
FQ213835.1
-
4
446
FQ213894.1
-
4
445
FQ215242.1
-
4
445
FQ215292.1
-
4
446
FQ216063.1
-
4
445
FQ216085.1
-
4
445
FQ216130.1
-
4
445
FQ216245.1
-
4
445
FQ216788.1
-
4
447
FQ216997.1
-
4
445
FQ217664.1
-
4
445
FQ219586.1
-
4
445
FQ219785.1
-
4
448
FQ220012.1
-
4
445
FQ220017.1
-
4
443
FQ220075.1
-
4
446
FQ220084.1
-
4
448
FQ220126.1
-
4
445
FQ220134.1
-
4
445
FQ220171.1
-
4
444
FQ220184.1
-
4
445
FQ220347.1
-
4
447
FQ220461.1
-
4
426
FQ220468.1
-
4
445
FQ220661.1
-
4
448
FQ220822.1
-
4
445
FQ220904.1
-
4
446
FQ220964.1
-
4
445
FQ221103.1
-
4
445
FQ221334.1
-
4
465
FQ221443.1
-
4
446
FQ221672.1
-
4
445
FQ222093.1
-
4
447
FQ222280.1
-
4
445
FQ222542.1
-
4
447
FQ222757.1
-
4
443
FQ223223.1
-
4
445
FQ224747.1
-
4
445
FQ225510.1
-
4
446
FQ226054.1
-
4
445
FQ226408.1
-
4
445
FQ227056.1
-
4
445
FQ227293.1
-
4
445
FQ228002.1
-
4
445
FQ228212.1
-
4
446
FQ229290.1
-
4
447
FQ229305.1
-
4
447
FQ229367.1
-
4
446
FQ229516.1
-
4
445
FQ229579.1
-
4
445
FQ229598.1
-
4
446
FQ229712.1
-
4
445
FQ229787.1
-
4
445
FQ229850.1
-
4
447
FQ229859.1
-
4
443
FQ229890.1
-
4
445
FQ229918.1
-
4
465
FQ229945.1
-
4
445
FQ229970.1
-
4
445
FQ230088.1
-
4
445
FQ230106.1
-
4
445
FQ230180.1
-
4
446
FQ230258.1
-
4
444
FQ231469.1
-
3
262
FQ231622.1
-
4
443
FQ232305.1
-
4
447
FQ232568.1
-
4
446
FQ233068.1
-
4
445
FQ233112.1
-
4
445
FQ233340.1
-
4
447
FQ233547.1
-
4
446
FQ233919.1
-
4
445
FQ234511.1
-
4
445
FQ234742.1
-
4
445
FQ234765.1
-
4
445
FQ234772.1
-
4
445
Back To Top
More Information
Panther Classification:
Molecular Function -
ribosomal protein
Gene Ontology Categories:
Function(s)
Process(es)
ribosomal small subunit assembly
endonucleolytic cleavage in ITS1 to separate SSU-rRNA from 5.8S rRNA and LSU-rRNA from tricistronic rRNA transcript (SSU-rRNA, 5.8S rRNA, LSU-rRNA)
endonucleolytic cleavage to generate mature 3'-end of SSU-rRNA from (SSU-rRNA, 5.8S rRNA, LSU-rRNA)
rRNA export from nucleus
translation
single organismal cell-cell adhesion
epithelial cell differentiation
structural constituent of ribosome
laminin receptor activity
structural molecule activity
ribosome binding
laminin binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-7745c474b-b52np:80/100.66.77.222:80.
git-commit: c3c9db9cc82507548abdd0ae6a4ade3f81f66292
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.20.0-2024.11.45-1.0