Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Car4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019174.3 | NP_062047.1 | 3 | 358 | ||
GenBank mRNA | BC097329.1 | - | 3 | 253 | 7445156 | |
FM062617.1 | - | 3 | 358 | |||
FQ222059.1 | - | 308 | ||||
S68245.1 | - | 3 | 301 |