Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019183.1 | NP_062056.1 | 6 | 1061 | ||
GenBank mRNA | BC127451.1 | - | 6 | 936 | 7379877 | |
CF109672.1 | - | 6 | 915 | |||
X80130.1 | - | 6 | 947 |
Gene Symbol: Acta2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | X00306.1 | - | 19 |